Methyl-2-aminoisonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-aminopyridine-4-carboxylate, Methyl 2-aminoisonicotinate

A118976-25G A118976-1G A118976-5G A118976-100G A118976-10G A118976-500G
103799-144EA 7 USD
103799-144 77108-450 77311-810 77311-812 77610-984 77610-986 77610-988
Methyl-2-aminoisonicotinate 98%
Methyl 2-aminoisonicotinate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 296.1 °C (760 mm Hg)
Melting Pt: 146…148 °C
Flash Pt: 132.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04039316
CAS Number: 6937-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR