Methyl-2-amino-6-fluoro-3-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-6-fluoro-3-nitrobenzoate

A109188-10G A109188-250MG A109188-1G
103797-464EA 30 USD
103797-464 103823-256 76809-852 76809-854 77310-078
Methyl-2-amino-6-fluoro-3-nitrobenzoate 97%
Methyl-2-amino-6-fluoro-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₇FN₂O₄
MW: 214.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839221
CAS Number: 346691-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR