Methyl 2-amino-6-bromobenzoate 98%

Supplier: Ambeed
Warning

A115541-100MG A115541-250MG
103798-586EA 33 USD
103798-586 103824-444 76810-152 76810-154
Methyl 2-amino-6-bromobenzoate 98%
Methyl 2-amino-6-bromobenzoate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
Boiling Pt: 297.2 °C (760 mm Hg)
Flash Pt: 133.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11977360
CAS Number: 135484-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR