Methyl-2-amino-5-nitrobenzoate 98%

Supplier: Ambeed
Warning

A156282-100MG A156282-250MG A156282-25G A156282-1G A156282-5G A156282-10G
103804-854EA 12 USD
103804-854 103843-056 76798-818 76798-820 77300-174 77318-812 77318-814 77592-922
Methyl-2-amino-5-nitrobenzoate 98%
Methyl-2-amino-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₈N₂O₄
Boiling Pt: 377.9 °C (760 mm Hg)
Flash Pt: 182.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00278157
CAS Number: 3816-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR