Methyl-2-amino-5-methoxybenzoate 97%
Supplier: Ambeed
|
Warning
|
A296548-100G
A296548-10G
A296548-1G
A296548-5G
A296548-25G
103814-102EA
7
USD
103814-102
103835-244
76787-642
76787-644
76837-916
76837-918
77305-694
Methyl-2-amino-5-methoxybenzoate 97%
Methyl-2-amino-5-methoxybenzoate
Precaución: for Research Use Only
Formula:
C₉H₁₁NO₃ Melting Pt: 37…38 °C Flash Pt: 145.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD09038482 CAS Number: 2475-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...