Methyl-2-amino-5-hydroxybenzoate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxyanthranilic acid methyll ester, Methyl 5-Hydroxyanthranilate, Methyl 2-amino-5-hydroxybenzoate, Methyl 2-Amino-5-hydroxybenzoate, 2-Amino-5-hydroxybenzoic acid methyll ester

A126999-100G A126999-250MG A126999-1G A126999-10G
103800-896EA 6 USD
103800-896 76782-082 76782-084 77313-144 77666-812
Methyl-2-amino-5-hydroxybenzoate 97%
Methyl-2-amino-5-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.1600 g/mol
Boiling Pt: 357.4 °C (760 mm Hg)
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD05863263
CAS Number: 1882-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR