Methyl-2-amino-5-fluorobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-Amino-5-fluorobenzoate, 5-Fluoroanthranilic acid methyll ester, Methyl 5-Fluoroanthranilate, 2-Amino-5-fluorobenzoic acid methyll ester, Methyl 2-amino-5-fluorobenzoate
A130738-100G
A130738-10G
A130738-1G
A130738-25G
A130738-5G
103801-470EA
11
USD
103801-470
77246-518
77246-520
77313-926
77313-928
77313-930
Methyl-2-amino-5-fluorobenzoate 98%
Methyl-2-amino-5-fluorobenzoate
Precaución: for Research Use Only
Formula:
C₈H₈FNO₂ MW: 169.16 g/mol Boiling Pt: 265.3 °C (760 mm Hg) Melting Pt: 34…37 °C Flash Pt: 132 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03791308 CAS Number: 319-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...