Methyl-2-amino-5-bromopyrimidine-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-5-bromopyrimidine-4-carboxylate

A296600-1G A296600-250MG
103814-106EA 144 USD
103814-106 103835-248 76837-924 76977-660
Methyl-2-amino-5-bromopyrimidine-4-carboxylate 95%
Methyl-2-amino-5-bromopyrimidine-4-carboxylate

Precaución: For research use only

Formula: C₆H₆BrN₃O₂
MW: 232.0400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09966084
CAS Number: 1034737-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR