Methyl-2-amino-5-bromo-3-methylbenzenecarboxylate 97%

Supplier: Ambeed
Warning

A313521-250MG A313521-1G
103825-008EA 17 USD
103825-008 103835-920 77015-184 77306-092
Methyl-2-amino-5-bromo-3-methylbenzenecarboxylate 97%
Methyl-2-amino-5-bromo-3-methylbenzenecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
Boiling Pt: 307.3±37.0 °C (760 mm Hg)
Melting Pt: 56…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06797379
CAS Number: 206548-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR