Methyl-2-amino-5-(trifluoromethyl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-5-(trifluoromethyl)benzoate

A165034-100G A165034-10G A165034-1G A165034-5G
103805-934EA 12 USD
103805-934 103844-192 76799-158 76799-160 77320-384 77320-386
Methyl-2-amino-5-(trifluoromethyl)benzoate 98%
Methyl-2-amino-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
Flash Pt: 114.632 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08234902
CAS Number: 117324-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR