Methyl 2-amino-4-methoxybenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-4-methoxylbenzoate

A443634-1G A443634-5G A443634-100G A443634-10G A443634-250MG A443634-25G
103827-698EA 13 USD
103827-698 103838-768 76842-544 76842-546 77168-818 77168-820 77168-822 77307-322
Methyl 2-amino-4-methoxybenzoate 97%
Methyl 2-amino-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.189 g/mol
Boiling Pt: 313.7 °C (760 mm Hg)
Flash Pt: 158.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06658470
CAS Number: 50413-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR