Methyl 2-amino-4-iodobenzoate 97%

Supplier: Ambeed
Warning

A128451-10G A128451-250MG A128451-1G A128451-5G
103801-124EA 47 USD
103801-124 76782-158 76782-160 77313-428 77313-430
Methyl 2-amino-4-iodobenzoate 97%
Methyl 2-amino-4-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₈INO₂
Storage Temperature: Refrigerator
MDL Number: MFCD13182440
CAS Number: 144550-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR