Methyl 2-amino-4-hydroxybenzoate 98%
Supplier: Ambeed
|
Warning
|
A148171-100MG
A148171-250MG
A148171-1G
103803-888EA
74
USD
103803-888
103842-002
77010-646
77010-648
77317-434
Methyl 2-amino-4-hydroxybenzoate 98%
Methyl 2-amino-4-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₈H₉NO₃ MW: 167.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08274816 CAS Number: 401568-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...