Methyl 2-amino-4-bromo-5-fluorobenzoate 95%

Supplier: Ambeed
Warning

A132703-250MG A132703-1G A132703-5G
103801-764EA 43 USD
103801-764 77095-026 77314-354 77314-356
Methyl 2-amino-4-bromo-5-fluorobenzoate 95%
Methyl 2-amino-4-bromo-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD22565660
CAS Number: 1395493-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR