Methyl 2-amino-4,6-dimethoxybenzoate 95%

Supplier: Ambeed
Warning

A710975-100MG A710975-250MG
103840-698EA 200 USD
103840-698 76992-758 76992-760
Methyl 2-amino-4,6-dimethoxybenzoate 95%
Methyl 2-amino-4,6-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄
MW: 211.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09031994
CAS Number: 379228-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR