Methyl 2-amino-4,6-difluorobenzoate 98%

Supplier: Ambeed
Warning

A142758-250MG A142758-1G
103803-210EA 110 USD
103803-210 76984-280 77299-308
Methyl 2-amino-4,6-difluorobenzoate 98%
Methyl 2-amino-4,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇F₂NO₂
MW: 187.143 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12159819
CAS Number: 379228-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR