Methyl-2-amino-4-(trifluoromethyl)benzoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-4-(trifluoromethyl)benzoate, 2-Amino-4-(trifluoromethyl)benzoic acid methyll ester

A320621-1G A320621-10G A320621-25G A320621-5G
103825-238EA 8 USD
103825-238 103836-172 76839-054 77114-714 77306-210 77306-212
Methyl-2-amino-4-(trifluoromethyl)benzoate 95%
Methyl-2-amino-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.1605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08543939
CAS Number: 61500-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR