Methyl-2-amino-3-methoxybenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-amino-3-methoxybenzoate
A113234-10G
A113234-250MG
A113234-25G
A113234-1G
A113234-5G
103798-200EA
11
USD
103798-200
103824-038
76970-994
76970-996
76970-998
77297-234
77310-860
Methyl-2-amino-3-methoxybenzoate 98%
Methyl-2-amino-3-methoxybenzoate
Precaución: for Research Use Only
Formula:
C₉H₁₁NO₃ MW: 181.19 g/mol Boiling Pt: 286.1 °C (760 mm Hg) Flash Pt: 133.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD11113109 CAS Number: 5121-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...