Methyl-2-amino-3-bromobenzoate 98%

Supplier: Ambeed
Warning

A100061-100G A100061-10G A100061-250MG A100061-25G A100061-5G A100061-1G
103795-226EA 6 USD
103795-226 103820-948 76970-102 76970-104 76970-106 76970-108 77296-558 77282-418
Methyl-2-amino-3-bromobenzoate 98%
Methyl 2-amino-3-bromobenzoate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
Melting Pt: 47…48 °C
Storage Temperature: Ambient
MDL Number: MFCD11042862
CAS Number: 104670-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR