Methyl-2-amino-3-(4-chlorophenyl)propanoate hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A393140-5G
A393140-1G
A393140-10G
103826-918EA
23
USD
103826-918
103837-958
76841-362
77307-026
77626-796
Methyl-2-amino-3-(4-chlorophenyl)propanoate hydrochloride 98%
Methyl-2-amino-3-(4-chlorophenyl)propanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₂ClNO₂·HCl Melting Pt: 186…189 °C Storage Temperature: Freezer |
MDL Number:
MFCD00012530 CAS Number: 14173-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...