Methyl 2-amino-2-(tetrahydro-2H-pyran-4-yl)acetate hydrochloride 95%

Supplier: Ambeed
Warning

A117215-100MG A117215-250MG A117215-1G
103798-852EA 291 USD
103798-852 103824-724 77161-068 77161-070 77297-522
Methyl 2-amino-2-(tetrahydro-2H-pyran-4-yl)acetate hydrochloride 95%
Methyl 2-amino-2-(tetrahydro-2H-pyran-4-yl)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClNO₃
MW: 209.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04037018
CAS Number: 1260637-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR