Methyl-2,6-dihydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,6-dihydroxybenzoate, 2,6-Dihydroxybenzoic acid methyll ester, gamma-Resorcylic acid methyll ester

A133291-100G A133291-100MG A133291-250MG A133291-1G A133291-25G A133291-5G
103801-856EA 8 USD
103801-856 76972-210 76972-212 76972-214 77314-492 77314-494 77314-496
Methyl-2,6-dihydroxybenzoate 98%
Methyl-2,6-dihydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 253.6 °C (760 mm Hg)
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00059619
CAS Number: 2150-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR