Methyl-2,6-difluorobenzoate 98%

Supplier: Ambeed
Warning

A319456-100G A319456-10G A319456-500G A319456-25G A319456-5G
103836-132EA 6 USD
103836-132 76802-876 76802-878 76802-880 76838-982 76838-984
Methyl-2,6-difluorobenzoate 98%
Methyl-2,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
Boiling Pt: 203…204 °C (760 mm Hg)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00051778
CAS Number: 13671-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR