Methyl-2,6-difluoro-3-nitrobenzoate 95%

Supplier: Ambeed
Warning

A183433-100G A183433-10G A183433-250MG A183433-1G A183433-25G A183433-5G
103815-642EA 9 USD
103815-642 103846-204 76872-832 76872-834 76872-836 77083-808 77083-810 77083-812
Methyl-2,6-difluoro-3-nitrobenzoate 95%
Methyl-2,6-difluoro-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₅F₂NO₄
Boiling Pt: 318.3 °C (760 mm Hg)
Flash Pt: 146.3 °C
Storage Temperature: Ambient
MDL Number: MFCD07368850
CAS Number: 84832-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR