Methyl 2,6-dibromoisonicotinate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,6-dibromopyridine-4-carboxylate, 2,6-Dibromo-4-(methoxycarbonyl)pyridine

A265226-1G A265226-5G A265226-250MG A265226-25G
103812-714EA 20 USD
103812-714 103852-608 77092-276 77092-278 77153-534 77153-536
Methyl 2,6-dibromoisonicotinate 96%
Methyl 2,6-dibromoisonicotinate

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₂
MW: 294.928 g/mol
Boiling Pt: 320.9±37.0 °C (760 mm Hg)
Flash Pt: 147.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11042543
CAS Number: 119308-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR