Methyl-2,5-dichloronicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,5-dichloronicotinate

A183941-100MG A183941-10G A183941-250MG A183941-1G
103815-692EA 8 USD
103815-692 103846-254 76799-818 76799-820 76799-822 77083-868
Methyl-2,5-dichloronicotinate 98%
Methyl-2,5-dichloronicotinate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Boiling Pt: 266.7 °C (760 mm Hg)
Melting Pt: 38…40 °C
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD03844857
CAS Number: 67754-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR