Methyl-2,5-dichloroisonicotinate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,5-dichloroisonicotinate, Methyl 2,5-dichloropyridine-4-carboxylate

A249802-100MG A249802-10G A249802-250MG A249802-25G
103811-892EA 23 USD
103811-892 103851-730 77000-654 77000-656 77000-658 77091-078
Methyl-2,5-dichloroisonicotinate 95%
Methyl 2,5-dichloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.0300 g/mol
Boiling Pt: 275.2 °C (760 mm Hg)
Melting Pt: 28…29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09951944
CAS Number: 623585-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR