Methyl-2,5-dibromobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,5-dibromobenzoate

A141808-10G A141808-1G A141808-5G
103803-066EA 11 USD
103803-066 76798-336 77299-250 77316-258
Methyl-2,5-dibromobenzoate 98%
Methyl 2,5-dibromobenzoate

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
MW: 293.94 g/mol
Boiling Pt: 291.3 °C (760 mm Hg)
Melting Pt: 48…51 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00144757
CAS Number: 57381-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR