Methyl-2,4-difluoro-5-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,4-difluoro-5-nitrobenzoate

A408536-25G A408536-100G A408536-1G
103827-186EA 9 USD
103827-186 103838-232 76841-746 77252-202 77307-128
Methyl-2,4-difluoro-5-nitrobenzoate 98%
Methyl 2,4-difluoro-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₅F₂NO₄
Storage Temperature: Ambient
MDL Number: MFCD07779369
CAS Number: 125568-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR