Methyl-2,4-dichloropyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A109465-100G A109465-10G A109465-25G A109465-1G A109465-5G
103797-510EA 14 USD
103797-510 103823-306 76718-004 76718-006 76718-008 77310-124 77310-126
Methyl-2,4-dichloropyrimidine-5-carboxylate 97%
Methyl-2,4-dichloropyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂
Boiling Pt: 299.3 °C (760 mm Hg)
Melting Pt: 30…34 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09910342
CAS Number: 3177-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR