Methyl-2,3-dichlorobenzoate 95%

Supplier: Ambeed
Warning

A241910-100G A241910-1G A241910-25G A241910-5G
103811-438EA 11 USD
103811-438 103851-246 76801-354 77090-402 77090-404 77090-406
Methyl-2,3-dichlorobenzoate 95%
Methyl-2,3-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
Boiling Pt: 270 °C (760 mm Hg)
Melting Pt: 33…38 °C
Flash Pt: 116.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00061099
CAS Number: 2905-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR