Methyl-2,3-diaminobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,3-diaminobenzoate

A151983-100MG A151983-250MG A151983-1G A151983-5G
103804-332EA 24 USD
103804-332 103842-482 76798-674 76798-676 77318-078 77318-080
Methyl-2,3-diaminobenzoate 97%
Methyl 2,3-diaminobenzoate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038589
CAS Number: 107582-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR