Methyl 5-bromo-2,3-diaminobenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2,3-diamino-5-bromobenzoate
A123579-100MG
A123579-10G
A123579-250MG
A123579-1G
A123579-5G
103800-418EA
23
USD
103800-418
77161-474
77161-476
77161-478
77312-538
77312-540
Methyl 5-bromo-2,3-diaminobenzoate 97%
Methyl 5-bromo-2,3-diaminobenzoate
Precaución: for Research Use Only
Formula:
C₈H₉BrN₂O₂ MW: 245.073 g/mol Boiling Pt: 351.9±37.0 °C (760 mm Hg) Flash Pt: 166.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD20037660 CAS Number: 1248541-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...