Methyl dimethoxyacetate 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2,2-dimethoxyacetate
A536324-10G
A536324-25G
A536324-5G
103810-438EA
374
USD
103810-438
103839-716
76726-332
76843-742
76843-744
Methyl dimethoxyacetate 96%
Methyl dimethoxyacetate
Precaución: for Research Use Only
Formula:
C₅H₁₀O₄ Boiling Pt: 169 °C (760 mm Hg) Flash Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008484 CAS Number: 89-91-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...