Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate 98%

Supplier: Ambeed
Warning

A154470-100G A154470-10G A154470-25G A154470-5G
103804-642EA 5 USD
103804-642 103842-832 76973-256 76973-258 77300-062 77318-558 77318-560
Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate 98%
Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₆F₂O₄
Storage Temperature: Ambient
MDL Number: MFCD06203771
CAS Number: 773873-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR