Methyl 2,2-diethoxyacetimidate 95%

Supplier: Ambeed
Warning

A321636-100MG A321636-250MG A321636-25G A321636-5G A321636-1G
103825-282EA 52 USD
103825-282 77239-592 77239-594 77239-596 77239-598 77306-244
Methyl 2,2-diethoxyacetimidate 95%
Methyl 2,2-diethoxyacetimidate

Precaución: for Research Use Only

Formula: C₇H₁₅NO₃
MW: 161.2 g/mol
Boiling Pt: 159.0±50.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD12031316
CAS Number: 76742-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR