Methyl 2,2,2-trimethoxyacetate 95%

Supplier: Ambeed
Warning

A156359-100G A156359-10G A156359-5G A156359-1G A156359-25G
103804-862EA 28 USD
103804-862 103843-064 76812-472 76812-474 77300-176 77318-826 77318-828
Methyl 2,2,2-trimethoxyacetate 95%
Methyl 2,2,2-trimethoxyacetate

Precaución: for Research Use Only

Formula: C₆H₁₂O₅
MW: 164.156 g/mol
Boiling Pt: 189.2 °C (760 mm Hg)
Flash Pt: 82.2 °C
Storage Temperature: Ambient
MDL Number: MFCD09701200
CAS Number: 18370-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR