Methyl-2,2,2-trichloroacetimidate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2,2,2-trichloroacetimidate
A187442-10G
A187442-1G
A187442-5G
103816-014EA
5
USD
103816-014
103846-604
76974-706
77302-082
77302-084
Methyl-2,2,2-trichloroacetimidate 97%
Methyl-2,2,2-trichloroacetimidate
Precaución: for Research Use Only
Formula:
C₃H₄Cl₃NO MW: 176.421 g/mol Boiling Pt: 148.5 °C (760 mm Hg) Flash Pt: 74 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001759 CAS Number: 2533-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...