Methyl-2-(triphenylphosphoranylidene)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl (triphenylphosphoranylidene)acetate, Methyl 2-(triphenylphosphoranylidene)acetate, Methoxycarbonylmethylene-Triphenylphosphorane

A197326-100G A197326-10G A197326-1G A197326-500G A197326-5G A197326-1000G A197326-25G
103809-324EA 177 USD
103809-324 76800-226 76800-228 76800-230 76800-232 76800-234 77085-534 77085-536
Methyl-2-(triphenylphosphoranylidene)acetate 98%
Methyl-2-(triphenylphosphoranylidene)acetate

Precaución: for Research Use Only

Formula: C₂₁H₁₉O₂P
Boiling Pt: 479.1 °C (760 mm Hg)
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00008455
CAS Number: 2605-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR