Methyl-2-(trifluoromethyl)isonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(trifluoromethyl)isonicotinate

A407742-250MG A407742-1G
103827-158EA 91 USD
103827-158 103838-200 76817-950 76841-694
Methyl-2-(trifluoromethyl)isonicotinate 98%
Methyl-2-(trifluoromethyl)isonicotinate

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MW: 205.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698093
CAS Number: 588702-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR