Methyl tetrahydropyran-4-ylacetate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(tetrahydro-2H-pyran-4-yl)acetate

A372837-10G A372837-1G
103826-518EA 20 USD
103826-518 103837-536 76817-532 76840-798
Methyl tetrahydropyran-4-ylacetate 97%
Methyl tetrahydropyran-4-ylacetate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.2000 g/mol
Boiling Pt: 208.5 °C (760 mm Hg)
Flash Pt: 90.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09972087
CAS Number: 156002-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR