Methyl quinoxalin-6-ylacetate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(quinoxalin-6-yl)acetate

A169952-250MG A169952-1G
103806-430EA 170 USD
103806-430 103844-696 77163-502 77301-014
Methyl quinoxalin-6-ylacetate 97%
Methyl quinoxalin-6-ylacetate

Precaución: Sealed in dry, at room temperature.

Formula: C₁₁H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD19706065
CAS Number: 1233318-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR