Methyl pyridin-3-ylacetate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(pyridin-3-yl)acetate

A251752-250MG A251752-1G A251752-5G
103812-006EA 19 USD
103812-006 103851-846 76786-830 77091-236 77091-238
Methyl pyridin-3-ylacetate 95%
Methyl pyridin-3-ylacetate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023617
CAS Number: 39998-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR