Methyl-2-pyridylacetate 97%

Supplier: Ambeed
Warning

Synonyms: 2-Pyridineacetic Acid Methyl Ester, Methyl 2-Pyridineacetate, Methyl 2-(pyridin-2-yl)acetate, 2-Pyridylacetic Acid Methyl Ester

A165429-100G A165429-10G A165429-500G A165429-1G A165429-25G A165429-5G
103805-968EA 9 USD
103805-968 103844-222 76720-278 76720-280 76720-282 77320-428 77320-430 77320-432
Methyl-2-pyridylacetate 97%
Methyl-2-pyridylacetate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.17 g/mol
Boiling Pt: 302.4 °C (760 mm Hg)
Flash Pt: 83.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00006358
CAS Number: 1658-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR