Methyl-2-(hydroxymethyl)acrylate 92% stabilized
Supplier: Ambeed
|
Danger
|
Stabilization:
Stabilised with hydroquinone (HQ)
A492654-250MG
A492654-10G
103828-230EA
10
USD
103828-230
103839-306
76789-818
76789-820
77594-546
Methyl-2-(hydroxymethyl)acrylate 92% stabilized
Methyl-2-(hydroxymethyl)acrylate
Precaución: for Research Use Only
Formula:
C₅H₈O₃ Boiling Pt: 184.8 °C (760 mm Hg) Flash Pt: 90.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD12545894 CAS Number: 15484-46-5 UN: 2922 ADR: 8,6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...