Methyl (diethylphosphono)acetate 97%

Supplier: Ambeed
Warning

A169653-100G A169653-10G A169653-500G A169653-25G A169653-5G
103844-656EA 5 USD
103844-656 76720-440 76720-442 76720-444 77081-744 77300-988
Methyl (diethylphosphono)acetate 97%
Methyl (diethylphosphono)acetate

Precaución: for Research Use Only

Formula: C₇H₁₅O₅P
Boiling Pt: 270.2 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009081
CAS Number: 1067-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR