Methyl-2-(chlorosulfonyl)benzoate 97%

Supplier: Ambeed
Danger

A863719-1G A863719-5G A863719-250MG A863719-10G
103841-260EA 23 USD
103841-260 76845-526 76845-528 77244-554 77573-742
Methyl-2-(chlorosulfonyl)benzoate 97%
Methyl 2-(chlorosulphonyl)benzoate

Precaución: for Research Use Only

Formula: C₈H₇ClO₄S
Boiling Pt: 344.8 °C (760 mm Hg)
Melting Pt: 62…63 °C
Flash Pt: 162.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009797
CAS Number: 26638-43-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR