Methyl (chlorosulfonyl)acetate 95%

Supplier: Ambeed
Danger

Synonyms: Methyl 2-chlorosulphonylacetate, Methyl 2-(chlorosulphonyl)acetate

A162385-250MG A162385-1G A162385-25G A162385-5G A162385-10G
103805-610EA 23 USD
103805-610 103843-860 76799-058 76799-060 77319-896 77319-898 77319-900 77879-101
Methyl (chlorosulfonyl)acetate 95%
Methyl (chlorosulfonyl)acetate

Precaución: for Research Use Only

Formula: C₃H₅ClO₄S
MW: 172.587 g/mol
Boiling Pt: 228.392 °C (760 mm Hg)
Flash Pt: 91.928 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075558
CAS Number: 56146-83-9
UN: 3265
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR