Methyl 2-(chlorosulfonyl)-5-nitrobenzoate 95%

Supplier: Ambeed
Danger

A443893-10G A443893-250MG A443893-1G
103827-702EA 200 USD
103827-702 103838-770 77265-020 77265-022 77307-324
Methyl 2-(chlorosulfonyl)-5-nitrobenzoate 95%
Methyl 2-(chlorosulfonyl)-5-nitrobenzoate

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₈H₆ClNO₆S
MW: 279.65 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25970336
CAS Number: 1039020-81-9
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR