Methyl 2-(chlorosulfonyl)-3-methylbenzoate 97%, Ambeed.Inc

Supplier: Ambeed
Danger

A111702-250MG A111702-1G A111702-5G A111702-25G
103797-886EA 10 USD
103797-886 103823-704 77008-948 77310-518 77310-520 77683-022
Methyl 2-(chlorosulfonyl)-3-methylbenzoate 97%, Ambeed.Inc
Methyl 2-(chlorosulfonyl)-3-methylbenzoate

Precaución: For research use only.

Formula: C₉H₉ClO₄S
MW: 248.68 g/mol
Boiling Pt: 366.6±35.0 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD09030584
CAS Number: 126535-26-0
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR